Assay Details
| Gene Symbol: CHCHD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018553.4 | XP_004018602.1 | 2 - 3 | 267 | 59 | |
| XM_012100278.3 | XP_011955668.1 | 2 - 3 | 238 | 59 | ||
More Information
Set Membership: |
|

CHCHD4
Genomic Map