Assay Details
| Gene Symbol: C2CD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965092.1 | XP_027820893.1 | 16 - 17 | 1684 | 130 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2CD6| Gene Symbol: C2CD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965092.1 | XP_027820893.1 | 16 - 17 | 1684 | 130 | |
Set Membership: |
|