Assay Details
| Gene Symbol: NT5C3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007912.3 | XP_004007961.1 | 3 - 4 | 400 | 103 | |
| XM_004007913.4 | XP_004007962.1 | 4 - 5 | 319 | 103 | ||
| XM_004007914.4 | XP_004007963.1 | 4 - 5 | 460 | 103 | ||
| XM_027968633.1 | XP_027824434.1 | 5 - 6 | 291 | 103 | ||
More Information
Set Membership: |
|

NT5C3A
Genomic Map