Assay Details
| Gene Symbol: PA2G4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006595.4 | XP_004006644.2 | 12 - 13 | 1312 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PA2G4| Gene Symbol: PA2G4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006595.4 | XP_004006644.2 | 12 - 13 | 1312 | 73 | |
Set Membership: |
|