Assay Details
| Gene Symbol: IPCEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012183166.3 | XP_012038556.2 | 5 - 6 | 606 | 84 | |
| XM_027972591.1 | XP_027828392.1 | 7 - 8 | 516 | 84 | ||
| XM_027972592.1 | XP_027828393.1 | 7 - 8 | 493 | 84 | ||
More Information
Set Membership: |
|

IPCEF1
Genomic Map