Assay Details
| Gene Symbol: CEBPZOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966926.1 | XP_027822727.1 | 2 - 3 | 152 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEBPZOS| Gene Symbol: CEBPZOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966926.1 | XP_027822727.1 | 2 - 3 | 152 | 102 | |
Set Membership: |
|