Assay Details
| Gene Symbol: CHST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962225.1 | XP_027818026.1 | 4 - 5 | 851 | 86 | |
| XM_027962226.1 | XP_027818027.1 | 5 - 6 | 1061 | 86 | ||
| XM_027962227.1 | XP_027818028.1 | 2 - 3 | 625 | 86 | ||
More Information
Set Membership: |
|

CHST3
Genomic Map