Assay Details
| Gene Symbol: LOC114111638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963449.1 | XP_027819250.1 | 5 - 6 | 1068 | 86 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC114111638| Gene Symbol: LOC114111638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963449.1 | XP_027819250.1 | 5 - 6 | 1068 | 86 | |