Assay Details
| Gene Symbol: CDNF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014237.4 | XP_004014286.1 | 2 - 3 | 374 | 103 | |
| XM_027976652.1 | XP_027832453.1 | 2 - 3 | 164 | 103 | ||
More Information
Set Membership: |
|

CDNF
Genomic Map