Assay Details
| Gene Symbol: RHBDD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020978.4 | XP_004021027.1 | 2 - 3 | 700 | 60 | |
| XM_012104476.3 | XP_011959866.1 | 3 - 4 | 836 | 60 | ||
| XM_015104213.2 | XP_014959699.1 | 4 - 5 | 795 | 60 | ||
| XM_027961303.1 | XP_027817104.1 | 3 - 4 | 1771 | 60 | ||
| XM_027961304.1 | XP_027817105.1 | 5 - 6 | 1170 | 60 | ||
| XM_027961305.1 | XP_027817106.1 | 4 - 5 | 958 | 60 | ||
| XM_027961306.1 | XP_027817107.1 | 6 - 7 | 999 | 60 | ||
| XM_027961307.1 | XP_027817108.1 | 5 - 6 | 979 | 60 | ||
More Information
Set Membership: |
|

RHBDD2
Genomic Map