Assay Details
| Gene Symbol: CCNQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012107098.2 | XP_011962488.1 | 4 - 5 | 592 | 64 | |
| XM_012107100.2 | XP_011962490.1 | 3 - 4 | 394 | 64 | ||
| XM_027962984.1 | XP_027818785.1 | 3 - 4 | 549 | 64 | ||
| XM_027962985.1 | XP_027818786.1 | 3 - 4 | 560 | 64 | ||
| XM_027962986.1 | XP_027818787.1 | 3 - 4 | 549 | 64 | ||
| XM_027962987.1 | XP_027818788.1 | 3 - 4 | 560 | 64 | ||
| XM_027962988.1 | XP_027818789.1 | 4 - 5 | 1203 | 64 | ||
| XM_027962989.1 | XP_027818790.1 | 3 - 4 | 513 | 64 | ||
More Information
Set Membership: |
|

CCNQ
Genomic Map