Assay Details
| Gene Symbol: SOX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967820.1 | XP_027823621.1 | 4 - 5 | 3331 | 65 | |
| XM_027967821.1 | XP_027823622.1 | 4 - 5 | 3226 | 65 | ||
More Information
Set Membership: |
|

SOX5
Genomic Map