Assay Details
| Gene Symbol: LOC101118600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005929.3 | XP_004005978.1 | 7 - 8 | 1186 | 114 | |
| XM_027966801.1 | XP_027822602.1 | 7 - 8 | 1186 | 114 | ||
More Information
Set Membership: |
|

LOC101118600
Genomic Map