Assay Details
| Gene Symbol: LOC101106554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019138.3 | XP_004019187.1 | 2 - 3 | 1965 | 95 | |
| XM_012101307.2 | XP_011956697.1 | 2 - 3 | 4064 | 95 | ||
More Information
Set Membership: |
|

LOC101106554
Genomic Map