Assay Details
| Gene Symbol: TRMT112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019654.3 | XP_004019703.2 | 2 - 3 | 231 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRMT112| Gene Symbol: TRMT112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019654.3 | XP_004019703.2 | 2 - 3 | 231 | 57 | |
Set Membership: |
|