Assay Details
| Gene Symbol: LOC114115347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971206.1 | XP_027827007.1 | 6 - 7 | 533 | 93 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC114115347| Gene Symbol: LOC114115347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971206.1 | XP_027827007.1 | 6 - 7 | 533 | 93 | |