Assay Details
| Gene Symbol: SNRNP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978597.1 | XP_027834398.1 | 7 - 8 | 715 | 65 | |
| XM_027978598.1 | XP_027834399.1 | 7 - 8 | 715 | 65 | ||
More Information
Set Membership: |
|

SNRNP70
Genomic Map