Assay Details
| Gene Symbol: CMSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002874.4 | XP_004002923.2 | 4 - 5 | 497 | 76 | |
| XM_012188621.2 | XP_012044011.2 | 4 - 5 | 345 | 76 | ||
| XM_015092406.2 | XP_014947892.2 | 4 - 5 | 312 | 76 | ||
| XM_027979289.1 | XP_027835090.1 | 4 - 5 | 533 | 76 | ||
| XM_027979310.1 | XP_027835111.1 | 4 - 5 | 560 | 76 | ||
More Information
Set Membership: |
|

CMSS1
Genomic Map