Assay Details
| Gene Symbol: LOC101103021 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019321.4 | XP_004019370.3 | 3 - 4 | 2000 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101103021| Gene Symbol: LOC101103021 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019321.4 | XP_004019370.3 | 3 - 4 | 2000 | 64 | |
Set Membership: |
|