Assay Details
| Gene Symbol: KCNN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002559.4 | XP_004002608.2 | 3 - 4 | 1832 | 69 | |
| XM_027975753.1 | XP_027831554.1 | 3 - 4 | 1832 | 69 | ||
| XM_027975761.1 | XP_027831562.1 | 3 - 4 | 1216 | 69 | ||
| XM_027975769.1 | XP_027831570.1 | 3 - 4 | 1089 | 69 | ||
More Information
Set Membership: |
|

KCNN3
Genomic Map