Assay Details
| Gene Symbol: GMNC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012097050.3 | XP_011952440.2 | 2 - 3 | 486 | 92 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: GMNC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012097050.3 | XP_011952440.2 | 2 - 3 | 486 | 92 | |