Assay Details
| Gene Symbol: CDCA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004001812.4 | XP_004001861.2 | 10 - 11 | 910 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDCA8| Gene Symbol: CDCA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004001812.4 | XP_004001861.2 | 10 - 11 | 910 | 63 | |
Set Membership: |
|