Assay Details
| Gene Symbol: MARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006543.4 | XP_004006592.2 | 11 - 12 | 1520 | 64 | |
| XM_012174672.2 | XP_012030062.2 | 9 - 10 | 1155 | 64 | ||
More Information
Set Membership: |
|

MARS1
Genomic Map