Assay Details
| Gene Symbol: LOC443240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968107.1 | XP_027823908.1 | 3 - 4 | 389 | 70 | |
| GenBank mRNA | EU016224.1 | - | 2 - 3 | 188 | 70 | |
More Information
Set Membership: |
|

LOC443240
Genomic Map