Assay Details
| Gene Symbol: GOPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011170.3 | XP_004011219.2 | 1 - 2 | 489 | 80 | |
| XM_004011171.4 | XP_004011220.2 | 1 - 2 | 492 | 80 | ||
More Information
Set Membership: |
|

GOPC
Genomic Map