Assay Details
| Gene Symbol: PPM1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005766.4 | XP_004005815.1 | 6 - 7 | 1173 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPM1G| Gene Symbol: PPM1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005766.4 | XP_004005815.1 | 6 - 7 | 1173 | 55 | |
Set Membership: |
|