Assay Details
| Gene Symbol: SPAG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977134.1 | XP_027832935.1 | 8 - 9 | 827 | 65 | |
| XM_027977135.1 | XP_027832936.1 | 8 - 9 | 558 | 65 | ||
More Information
Set Membership: |
|

SPAG4
Genomic Map