Assay Details
| Gene Symbol: HHLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097654.2 | XP_014953140.2 | 10 - 11 | 946 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HHLA1| Gene Symbol: HHLA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097654.2 | XP_014953140.2 | 10 - 11 | 946 | 67 | |
Set Membership: |
|