Assay Details
| Gene Symbol: CASP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012187488.3 | XP_012042878.2 | 6 - 7 | 1005 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CASP9| Gene Symbol: CASP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012187488.3 | XP_012042878.2 | 6 - 7 | 1005 | 62 | |
Set Membership: |
|