Assay Details
| Gene Symbol: CCDC162P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012182637.3 | XP_012038027.2 | 44 - 45 | 8133 | 98 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC162P| Gene Symbol: CCDC162P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012182637.3 | XP_012038027.2 | 44 - 45 | 8133 | 98 | |