Assay Details
| Gene Symbol: LOC101105041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020042.4 | XP_004020091.2 | 11 - 12 | 930 | 146 | |
| XM_012102578.3 | XP_011957968.2 | 11 - 12 | 808 | 146 | ||
| XM_027960170.1 | XP_027815971.1 | 10 - 11 | 1009 | 146 | ||
More Information
Set Membership: |
|

LOC101105041
Genomic Map