Assay Details
| Gene Symbol: ACTR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006649.4 | XP_004006698.1 | 1 - 2 | 121 | 78 | |
| XM_012174869.3 | XP_012030259.1 | 1 - 2 | 121 | 78 | ||
More Information
Set Membership: |
|

ACTR6
Genomic Map