Assay Details
| Gene Symbol: TRAF3IP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011198.4 | XP_004011247.1 | 4 - 5 | 1981 | 83 | |
| XM_015097341.2 | XP_014952827.1 | 4 - 5 | 1976 | 83 | ||
More Information
Set Membership: |
|

TRAF3IP2
Genomic Map