Assay Details
| Gene Symbol: PRKN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972639.1 | XP_027828440.1 | 9 - 10 | 1209 | 74 | |
| XM_027972640.1 | XP_027828441.1 | 9 - 10 | 1195 | 74 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PRKN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972639.1 | XP_027828440.1 | 9 - 10 | 1209 | 74 | |
| XM_027972640.1 | XP_027828441.1 | 9 - 10 | 1195 | 74 | ||