Assay Details
| Gene Symbol: AQP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010567.4 | XP_004010616.1 | 4 - 5 | 819 | 73 | |
| GenBank mRNA | AF548384.1 | - | 3 - 4 | 361 | 73 | |
More Information
Set Membership: |
|

AQP9
Genomic Map