Assay Details
| Gene Symbol: DALRD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012100083.2 | XP_011955473.2 | 7 - 8 | 1835 | 67 | |
| XM_027957662.1 | XP_027813463.1 | 7 - 8 | 1839 | 67 | ||
More Information
Set Membership: |
|

DALRD3
Genomic Map