Assay Details
Gene Symbol: C20H6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959034.1 | XP_027814835.1 | 8 - 9 | 1137 | 113 | |
XM_027959035.1 | XP_027814836.1 | 8 - 9 | 1369 | 113 | ||
XM_027959036.1 | XP_027814837.1 | 7 - 8 | 1079 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
