Assay Details
| Gene Symbol: SRPRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960315.1 | XP_027816116.1 | 6 - 7 | 932 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SRPRB| Gene Symbol: SRPRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960315.1 | XP_027816116.1 | 6 - 7 | 932 | 63 | |
Set Membership: |
|