Assay Details
| Gene Symbol: SLC66A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005688.4 | XP_004005737.2 | 2 - 3 | 319 | 71 | |
| XM_027966524.1 | XP_027822325.1 | 2 - 3 | 319 | 71 | ||
More Information
Set Membership: |
|

SLC66A3
Genomic Map