Assay Details
| Gene Symbol: SLC66A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005688.4 | XP_004005737.2 | 4 - 5 | 455 | 67 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC66A3| Gene Symbol: SLC66A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005688.4 | XP_004005737.2 | 4 - 5 | 455 | 67 | |