Assay Details
| Gene Symbol: C1H3orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002845.4 | XP_004002894.2 | 2 - 3 | 694 | 92 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1H3orf38| Gene Symbol: C1H3orf38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002845.4 | XP_004002894.2 | 2 - 3 | 694 | 92 | |