Assay Details
| Gene Symbol: WT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979592.1 | XP_027835393.1 | 8 - 9 | 860 | 75 | |
| XM_027979593.1 | XP_027835394.1 | 8 - 9 | 826 | 75 | ||
| XM_027979594.1 | XP_027835395.1 | 7 - 8 | 809 | 75 | ||
| XM_027979595.1 | XP_027835396.1 | 8 - 9 | 786 | 75 | ||
| XM_027979596.1 | XP_027835397.1 | 6 - 7 | 600 | 75 | ||
| GenBank mRNA | AY115591.1 | - | 7 - 8 | 616 | 75 | |
More Information
Set Membership: |
|

WT1
Genomic Map