Assay Details
| Gene Symbol: SHMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006554.3 | XP_004006603.1 | 7 - 8 | 946 | 63 | |
| XM_012174695.3 | XP_012030085.1 | 7 - 8 | 894 | 63 | ||
| XM_012174696.3 | XP_012030086.1 | 6 - 7 | 1132 | 63 | ||
| XM_027967563.1 | XP_027823364.1 | 7 - 8 | 908 | 63 | ||
| XM_027967564.1 | XP_027823365.1 | 6 - 7 | 846 | 63 | ||
More Information
Set Membership: |
|

SHMT2
Genomic Map