Assay Details
Gene Symbol: TPM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008453.3 | XP_004008502.1 | 3 - 4 | 414 | 70 | |
XM_004008454.3 | XP_004008503.1 | 3 - 4 | 420 | 70 | ||
XM_027969479.1 | XP_027825280.1 | 2 - 3 | 391 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
