Assay Details
| Gene Symbol: CRELD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018277.4 | XP_004018326.1 | 7 - 8 | 814 | 54 | |
| XM_012099504.2 | XP_011954894.1 | 7 - 8 | 836 | 54 | ||
More Information
Set Membership: |
|

CRELD1
Genomic Map