Assay Details
| Gene Symbol: MYLK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012189708.3 | XP_012045098.1 | 4 - 5 | 1846 | 61 | |
| XM_012189711.3 | XP_012045101.1 | 3 - 4 | 1021 | 61 | ||
| XM_012189713.3 | XP_012045103.1 | 3 - 4 | 1025 | 61 | ||
More Information
Set Membership: |
|

MYLK3
Genomic Map