Assay Details
| Gene Symbol: ALDH3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975464.1 | XP_027831265.1 | 7 - 8 | 1115 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALDH3A1| Gene Symbol: ALDH3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975464.1 | XP_027831265.1 | 7 - 8 | 1115 | 65 | |
Set Membership: |
|