Assay Details
| Gene Symbol: KIFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015102652.2 | XP_014958138.2 | 4 - 5 | 407 | 63 | |
| XM_015102653.2 | XP_014958139.2 | 4 - 5 | 423 | 63 | ||
More Information
Set Membership: |
|

KIFC1
Genomic Map