Assay Details
| Gene Symbol: LOC101106044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012162676.3 | XP_012018066.2 | 28 - 29 | 3999 | 76 | |
| XM_027970718.1 | XP_027826519.1 | 28 - 29 | 3999 | 76 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101106044| Gene Symbol: LOC101106044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012162676.3 | XP_012018066.2 | 28 - 29 | 3999 | 76 | |
| XM_027970718.1 | XP_027826519.1 | 28 - 29 | 3999 | 76 | ||