Assay Details
| Gene Symbol: NEDD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012174840.3 | XP_012030230.2 | 12 - 13 | 1760 | 63 | |
| XM_015094814.2 | XP_014950300.2 | 12 - 13 | 1731 | 63 | ||
| XM_015094815.2 | XP_014950301.1 | 10 - 11 | 1481 | 63 | ||
| XM_027967639.1 | XP_027823440.1 | 12 - 13 | 1858 | 63 | ||
More Information
Set Membership: |
|

NEDD1
Genomic Map